(R)-(+)-2,2'-Bis(di-i-propylphosphino)-6,6'-dimethoxy-1,1'-biphenyl ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-0654-100MG 15-0654-500MG
101418-602EA 140.12 USD
101418-602 101418-604
(R)-(+)-2,2'-Bis(di-i-propylphosphino)-6,6'-dimethoxy-1,1'-biphenyl ≥97%
(R)-(+)-2,2'-Bis(di-i-propylphosphino)-6,6'-dimethoxy-1,1'-biphenyl

BIPHEP, Phosphine

Formula: C₂₆H₄₀O₂P₂
MDL Number: MFCD09753011
CAS Number: 150971-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR