(R)-(-)-2,2'-Bis[di(3,5-di-i-propyl-4-dimethylaminophenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-0652-100MG 15-0652-500MG
101418-594EA 140.12 USD
101418-594 101418-596
(R)-(-)-2,2'-Bis[di(3,5-di-i-propyl-4-dimethylaminophenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl ≥97%
(R)-(-)-2,2'-Bis[di(3,5-di-i-propyl-4-dimethylaminophenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl

BIPHEP, Phosphine

Formula: C₇₀H₁₀₀N₄O₂P₂
MW: 1091.5100 g/mol
MDL Number: MFCD09753010
CAS Number: 352655-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR