6,6'-Bis(diphenylphosphino)-1,1',3,3'-tetrahydro[5,5']biisobenzofuran ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

15-0504-100MG 15-0504-500MG
101442-248EA 107.57 USD
101442-248 101442-250
6,6'-Bis(diphenylphosphino)-1,1',3,3'-tetrahydro[5,5']biisobenzofuran ≥99%
6,6'-Bis(diphenylphosphino)-1,1',3,3'-tetrahydro[5,5']biisobenzofuran

Phosphine

MDL Number: MFCD10565624
CAS Number: 959864-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR