S-(-)-6,6'-Bis(diphenylphosphino)-2,2',3,3'-tetrahydro-5,5'-bi-1,4-benzodioxin ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-SYNPHOS™

15-0491-500MG 15-0491-100MG
100209-692EA 471.32 USD
100209-692 100209-690
S-(-)-6,6'-Bis(diphenylphosphino)-2,2',3,3'-tetrahydro-5,5'-bi-1,4-benzodioxin ≥97%
S-(-)-6,6'-Bis(diphenylphosphino)-2,2',3,3'-tetrahydro-5,5'-bi-1,4-benzodioxin

Phosphine

MDL Number: MFCD05861608
CAS Number: 503538-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR