(R)-(+)-2,2'-Bis[di(3,5-xylyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-0488-100MG 15-0488-500MG
101418-586EA 140.12 USD
101418-586 101418-588
(R)-(+)-2,2'-Bis[di(3,5-xylyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl ≥97%
(R)-(+)-2,2'-Bis[di(3,5-xylyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl

BIPHEP, Phosphine

Formula: C₄₆H₄₈O₂P₂
MW: 694.8197 g/mol
MDL Number: MFCD09753009
CAS Number: 394248-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR