S-(+)-5,5'-Bis(diphenylphosphino)-2,2,2',2'-tetrafluoro-4,4'-bi-1,3-benzodioxole, dichloromethane adduct ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-0487-500MG 15-0487-100MG
100203-972EA 819.51 USD
100203-972 100203-970
S-(+)-5,5'-Bis(diphenylphosphino)-2,2,2',2'-tetrafluoro-4,4'-bi-1,3-benzodioxole, dichloromethane adduct ≥97%
S-(+)-5,5'-Bis(diphenylphosphino)-2,2,2',2'-tetrafluoro-4,4'-bi-1,3-benzodioxole, dichloromethane adduct

Phosphine

MDL Number: MFCD05861607
CAS Number: 503538-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR