Di(1-adamantyl)-n-butylphosphine ≥95%
Supplier: STREM CHEMICALS INC MS
Synonyms:
n-butyl-di-adamantylphosphine, [cataCXium® A], Bis(1-adamantyl)-butyl-phosphane, Butyldi-1-adamantylphosphine, Di(adamantan-1-yl)(butyl)phosphine
15-0483-5G
15-0483-1G
100209-684EA
467.08
USD
100209-684
100209-682
Di(1-adamantyl)-n-butylphosphine ≥95%
Di(1-adamantyl)-n-butylphosphine
cataCXium, Phosphine
Formula:
C₂₄H₃₉P MW: 358.55 g/mol Melting Pt: 100 °C |
MDL Number:
MFCD05861606 CAS Number: 321921-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...