1,2-Bis[(2S,5S)-2,5-diphenyl-1-phospholanyl]ethane ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (+)-1,2-Bis((2S,5S)-2,5-diphenylphospholano)ethane, 1,2-Bis((2S,5S)-2,5-diphenylphospholan-1-yl)ethane, (S,S)-Ph-BPE

15-0474-100MG 15-0474-2G 15-0474-500MG
101324-578EA 148.62 USD
101324-578 101324-582 101324-580
1,2-Bis[(2S,5S)-2,5-diphenyl-1-phospholanyl]ethane ≥98%
1,2-Bis[(2S,5S)-2,5-diphenyl-1-phospholanyl]ethane

BPE, DUPHOS, Phosphine

Formula: C₃₄H₃₆P₂
MW: 506.61 g/mol
Melting Pt: 144 °C
MDL Number: MFCD07369028
CAS Number: 824395-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR