(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R,R)-Ph-BPE, 1,2-Bis((2R,5R)-2,5-diphenylphospholan-1-yl)ethane

15-0473-500MG 15-0473-100MG 15-0473-2G
101324-574EA 564.74 USD
101324-574 101324-572 101324-576
(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane ≥95%
(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane

BPE, DUPHOS, Phosphine

Formula: C₃₄H₃₆P₂
MW: 506.61 g/mol
Melting Pt: 144 °C
MDL Number: MFCD07369027
CAS Number: 528565-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR