Bis(2-methoxyphenyl)phenylphosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(2-methoxyphenyl)(phenyl)phosphine

15-0466-25G 15-0466-1G 15-0466-5G
100201-456EA 390.65 USD
100201-456 100209-674 100209-676
Bis(2-methoxyphenyl)phenylphosphine ≥98%
Bis(2-methoxyphenyl)phenylphosphine

Phosphine

Formula: C₂₀H₁₉O₂P
MW: 322.337 g/mol
Melting Pt: 163…164 °C
MDL Number: MFCD00061478
CAS Number: 36802-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR