1,2-Bis(phosphino)benzene ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

15-0461-1G 15-0461-5G
100206-328EA 270.34 USD
100206-328 100197-994
1,2-Bis(phosphino)benzene ≥98%
1,2-Bis(phosphino)benzene

Phosphine

Boiling Pt: 53…55 °C (0.25 mmHg)
Density: 1,101
MDL Number: MFCD00142463
CAS Number: 80510-04-9
UN: 2845
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR