1,2-Bis(phosphino)ethane ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

15-0459-250MG 15-0459-1G
100206-326EA 175.51 USD
100206-326 100210-726
1,2-Bis(phosphino)ethane ≥99%
1,2-Bis(phosphino)ethane

Phosphine

Formula: C₂H₈P₂
Boiling Pt: 109…110 °C
MDL Number: MFCD00049195
CAS Number: 5518-62-7
UN: 2845
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR