Bis(triphenylphosphoranylidene)ammonium chloride ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1,1,1-Triphenyl-N-(triphenylphosphoranylidene)phosphoraniminium chloride, Bis(triphenylphosphine)iminium chloride

15-0455-25G 15-0455-5G
100200-244EA 329.78 USD
100200-244 100204-928
Bis(triphenylphosphoranylidene)ammonium chloride ≥97%
Bis(triphenylphosphoranylidene)ammonium chloride

Phosphine

Formula: C₃₆H₃₀ClNP₂
MW: 574.041 g/mol
Melting Pt: 260…265 °C
MDL Number: MFCD00151523
CAS Number: 21050-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR