1,3-Bis(diphenylphosphino)propane monooxide ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-0449-1G 15-0449-5G
100201-452EA 120.31 USD
100201-452 100201-454
1,3-Bis(diphenylphosphino)propane monooxide ≥97%
1,3-Bis(diphenylphosphino)propane monooxide

Phosphine

MDL Number: MFCD03412077
CAS Number: 85685-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR