Bis(diisopropylamino)chlorophosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Bis(di-i-propylamino)chlorophosphine

15-0448-5G 15-0448-25G
100211-008EA 200.98 USD
100211-008 100209-472
Bis(diisopropylamino)chlorophosphine ≥97%
Bis(diisopropylamino)chlorophosphine

Phosphine

Formula: C₁₂H₂₈ClN₂P
MW: 266.79 g/mol
MDL Number: MFCD00061482
CAS Number: 56183-63-2
UN: 1759
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR