(R)-(+)-Bis-(1,2-diphenylphosphino)propane ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(R)-Propane-1,2-diylbis(diphenylphosphine), (R)-PROPHOS, R-(+)-1,2-Bis(diphenylphosphino)propane
15-0440-1G
15-0440-250MG
100211-006EA
486.89
USD
100211-006
100197-396
(R)-(+)-Bis-(1,2-diphenylphosphino)propane ≥99%
(R)-(+)-Bis-(1,2-diphenylphosphino)propane
Phosphine
Formula:
C₂₇H₂₆P₂ Melting Pt: 69…71 °C (sealed tube) |
MDL Number:
MFCD00012309 CAS Number: 67884-32-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...