4,6-Bis(diphenylphosphino)phenoxazine ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
4,6-Bis(diphenylphosphino)-10H-phenoxazine, NIXANTPHOS
15-0437-2G
15-0437-500MG
100200-638EA
440.18
USD
100200-638
100204-926
4,6-Bis(diphenylphosphino)phenoxazine ≥98%
4,6-Bis(diphenylphosphino)phenoxazine
Phosphine, Pincer Ligands and Complexes, XANTPHOS
Formula:
C₃₆H₂₇NOP₂ MW: 551.57 g/mol |
MDL Number:
MFCD03788937 CAS Number: 261733-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...