(2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (2S,4S)-Pentane-2,4-diylbis(diphenylphosphine), (S,S)-BDPP

15-0431-5G 15-0431-1G
100200-858EA 1167.69 USD
100200-858 100200-856
(2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane ≥99%
(2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane

Phosphine

Formula: C₂₉H₃₀P₂
MW: 440.50 g/mol
Melting Pt: 81 °C
MDL Number: MFCD00058923
CAS Number: 77876-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR