(R)-Phanephos ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-4,12-Bis(diphenylphosphino)-[2.2]paracyclophane, (R)-PHANEPHOS, (R)-(-)-4,12-Bis(diphenylphosphino)-[2.2]-paracyclophane

15-0425-100MG 15-0425-500MG
100196-066EA 159.94 USD
100196-066 100196-068
(R)-Phanephos ≥95%
(R)-Phanephos

PHANEPHOS, Phosphine

Formula: C₄₀H₃₄P₂
MW: 576.646 g/mol
Melting Pt: 222…225 °C
MDL Number: MFCD03094575
CAS Number: 364732-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR