1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]benzene ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S,S)-Isopropyl-DuPhos, (-)-1,2-Bis((2S,5S)-2,5-di-i-propylphospholano)benzene, (S,S)-i-Pr-DUPHOS

15-0411-500MG 15-0411-100MG
100208-836EA 409.05 USD
100208-836 100208-834
1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]benzene ≥98%
1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]benzene

BPE, DUPHOS, Phosphine

Formula: C₂₆H₄₄P₂
MW: 418.58 g/mol
Melting Pt: 40 °C
MDL Number: MFCD02684550
CAS Number: 147253-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR