1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]benzene ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1,2-Bis((2R,5R)-2,5-diisopropylphospholan-1-yl)benzene, (R,R)-i-Pr-DUPHOS, (+)-1,2-Bis((2R,5R)-2,5-di-i-propylphospholano)benzene

15-0410-500MG 15-0410-100MG 15-0410-2G
100208-832EA 493.97 USD
100208-832 100208-830 100201-188
1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]benzene ≥98%
1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]benzene

BPE, DUPHOS, Phosphine

Formula: C₂₆H₄₄P₂
MW: 418.586 g/mol
Melting Pt: 40 °C
MDL Number: MFCD02684549
CAS Number: 136705-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR