Bis(diphenylphosphino)methane ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
[(1,1-Diphenylphosphino)methyl](diphenyl)phosphine, DPPM
15-0400-25G
15-0400-1G
15-0400-5G
100205-916EA
273.17
USD
100205-916
100201-450
100198-614
Bis(diphenylphosphino)methane ≥97%
Bis(diphenylphosphino)methane
Phosphine
Formula:
C₂₅H₂₂P₂ Melting Pt: 116…119 °C |
MDL Number:
MFCD00003537 CAS Number: 2071-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...