Bis(2-diphenylphosphinoethyl)phenylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: ((Phenylphosphinediyl)bis(ethane-2,1-diyl))bis(diphenylphosphine), TRIPHOS

15-0350-5G 15-0350-1G
100198-612EA 351.02 USD
100198-612 100198-610
Bis(2-diphenylphosphinoethyl)phenylphosphine ≥97%
Bis(2-diphenylphosphinoethyl)phenylphosphine

Phosphine, Pincer Ligands and Complexes

Formula: C₃₄H₃₃P₃
MW: 534.559 g/mol
Melting Pt: 130…134 °C
MDL Number: MFCD00003048
CAS Number: 23582-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR