1,2-Bis(diphenylphosphino)ethane monooxide ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-0205-1G 15-0205-5G
100199-764EA 111.82 USD
100199-764 100199-766
1,2-Bis(diphenylphosphino)ethane monooxide ≥97%
1,2-Bis(diphenylphosphino)ethane monooxide

Phosphine

MDL Number: MFCD00345406
CAS Number: 984-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR