(R)-(+)-2,2'-Bis(diphenylphosphino)-6,6'-dimethoxy-1,1'-biphenyl ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-MeO-BIPHEP, (R)-(6,6'-Dimethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(diphenylphosphine)

15-0178-500MG 15-0178-100MG
101418-556EA 518.03 USD
101418-556 101418-554
(R)-(+)-2,2'-Bis(diphenylphosphino)-6,6'-dimethoxy-1,1'-biphenyl ≥97%
(R)-(+)-2,2'-Bis(diphenylphosphino)-6,6'-dimethoxy-1,1'-biphenyl

BIPHEP, Phosphine

Formula: C₃₈H₃₂O₂P₂
MW: 582.6100 g/mol
MDL Number: MFCD03095430
CAS Number: 133545-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR