(S)-(+)-1,13-Bis(diphenylphosphino)-7,8-dihydro-6H-dibenzo[f,h][1,5]dioxonin ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-1,13-Bis(diphenylphosphino)-7,8-dihydro-6H-dibenzo[f,h][1,5]dioxonine, (S)-C3-TUNEPHOS

15-0176-500MG 15-0176-100MG
101324-570EA 755.82 USD
101324-570 101324-568
(S)-(+)-1,13-Bis(diphenylphosphino)-7,8-dihydro-6H-dibenzo[f,h][1,5]dioxonin ≥95%
(S)-(+)-1,13-Bis(diphenylphosphino)-7,8-dihydro-6H-dibenzo[f,h][1,5]dioxonin

Phosphine

Formula: C₃₉H₃₂O₂P₂
MW: 594.6200 g/mol
MDL Number: MFCD06658115
CAS Number: 486429-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR