(S,S)-(-)-2,3-Bis(diphenylphosphino)butane

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (2S,3S)-(-)-Bis(diphenylphosphino)butane, (2S,3S)-Butane-2,3-diylbis(diphenylphosphine), (S,S)-CHIRAPHOS

15-0160-5G 15-0160-1G 15-0160-250MG
100203-738EA 1883.88 USD
100203-738 100203-736 100200-414
(S,S)-(-)-2,3-Bis(diphenylphosphino)butane
(S,S)-(-)-2,3-Bis(diphenylphosphino)butane

Phosphine

Formula: C₂₈H₂₈P₂
MW: 426.47 g/mol
Melting Pt: 108…109 °C
MDL Number: MFCD00009836
CAS Number: 64896-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR