(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-6,6'-dimethoxy-1,1'-biphenyl ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-0157-500MG 15-0157-100MG
101418-544EA 518.03 USD
101418-544 101418-542
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-6,6'-dimethoxy-1,1'-biphenyl ≥97%
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-6,6'-dimethoxy-1,1'-biphenyl

BIPHEP, Phosphine

Formula: C₄₂H₄₀O₂P₂
MDL Number: MFCD09753006
CAS Number: 133545-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR