1,2-Bis(diphenylphosphino)benzene ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

15-0130-5G 15-0130-1G
100202-944EA 468.49 USD
100202-944 100202-942
1,2-Bis(diphenylphosphino)benzene ≥98%
1,2-Bis(diphenylphosphino)benzene

Phosphine

Formula: C₃₀H₂₄P₂
Melting Pt: 185…187 °C
MDL Number: MFCD00014081
CAS Number: 13991-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR