N,N-Bis(diphenylphosphino)amine ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

15-0110-5G 15-0110-1G
100201-448EA 768.55 USD
100201-448 100201-446
N,N-Bis(diphenylphosphino)amine ≥98%
N,N-Bis(diphenylphosphino)amine

Phosphine

Melting Pt: 143…145 °C
MDL Number: MFCD00233855
CAS Number: 2960-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR