1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]ethane ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 5-(2-methylthiazol-4-yl)thiophene-3-carbonitrile, 5-(2-Methyl-1,3-thiazol-4-yl)-3-thiophenecarbonitrile, 4-(4-Cyano-2-thienyl)-2-methylthiazole, (S,S)-Me-BPE, (-)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane, (S,S)-Methyl-BPE

15-0105-100MG 15-0105-2G 15-0105-500MG
100208-826EA 108.98 USD
100208-826 100201-186 100208-828
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]ethane ≥98%
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]ethane

BPE, DUPHOS, Phosphine

Formula: C₁₄H₂₈P₂
MW: 258.326 g/mol
Boiling Pt: 64…67 °C (0.06 mmHg)
Density: 0,938
MDL Number: MFCD01073771
CAS Number: 136779-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR