1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]ethane ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (+)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)ethane, 1,2-Bis[(2R,5R)-2,5-dimethylphospholano]ethane, 1,2-Bis[(2R,5R)-2,5-dimethylphospholan-1-yl]ethane, (R,R)-Me-BPE

15-0104-500MG 15-0104-2G 15-0104-100MG
100208-824EA 401.97 USD
100208-824 100201-184 100208-822
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]ethane ≥98%
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]ethane

BPE, DUPHOS, Phosphine

Formula: C₁₄H₂₈P₂
MW: 258.326 g/mol
Boiling Pt: 64…67 °C (0.06 mmHg)
Density: 0,940
MDL Number: MFCD01073770
CAS Number: 129648-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR