(-)-1,2-Bis((2S,5S)-2,5-diethylphospholano)ethane ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S,S)-Et-BPE

15-0102-500MG 15-0102-100MG
100198-592EA 401.97 USD
100198-592 100198-590
(-)-1,2-Bis((2S,5S)-2,5-diethylphospholano)ethane ≥98%
(-)-1,2-Bis((2S,5S)-2,5-diethylphospholano)ethane

BPE, DUPHOS, Phosphine

Boiling Pt: 104…106 °C (0.05 mmHg)
MDL Number: MFCD01073769
CAS Number: 136779-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR