(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]ethyl]-2,5-diethylphospholane ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

15-0101-500MG 15-0101-100MG
100198-588EA 406.22 USD
100198-588 100198-586
(2R,5R)-1-[2-[(2R,5R)-2,5-diethylphospholan-1-yl]ethyl]-2,5-diethylphospholane ≥98%

BPE, DUPHOS, Phosphine

Formula: C₁₈H₃₆P₂
Boiling Pt: 104…106 °C (0,05 mmHg)
Density: 0,939
MDL Number: MFCD01073768
CAS Number: 136705-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR