Bis(diphenylphosphino)acetylene ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1,2-Bis(diphenylphosphino)ethyne

15-0100-1G 15-0100-5G
100197-388EA 131.63 USD
100197-388 100197-390
Bis(diphenylphosphino)acetylene ≥97%
Bis(diphenylphosphino)acetylene

Phosphine

Formula: C₂₆H₂₀P₂
MW: 394.394 g/mol
Melting Pt: 85…87 °C
MDL Number: MFCD00003045
CAS Number: 5112-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR