(+)-1,2-Bis[(2S,5S)-2,5-diethyl-1-phospholanyl]benzene ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene, 1,2-Bis((2S,5S)-2,5-diethylphospholan-1-yl)benzene, (S,S)-Et-DUPHOS

15-0098-100MG 15-0098-500MG 15-0098-2G
100208-818EA 118.89 USD
100208-818 100208-820 100201-182
(+)-1,2-Bis[(2S,5S)-2,5-diethyl-1-phospholanyl]benzene ≥98%
(+)-1,2-Bis[(2S,5S)-2,5-diethyl-1-phospholanyl]benzene

BPE, DUPHOS, Phosphine

Formula: C₂₂H₃₆P₂
MW: 362.478 g/mol
Density: 1.010
MDL Number: MFCD00142321
CAS Number: 136779-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR