1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene, 1,2-Bis((2R,5R)-2,5-dimethylphospholan-1-yl)benzene, 1,2-Bis[(2R,5R)-2,5-dimethylphospholan-1-yl]benzene, 1,2-Bis[(2R,5R)-2,5-dimethylphospholano]benzene, (R,R)-Me-DUPHOS

15-0096-500MG 15-0096-2G 15-0096-100MG
100208-812EA 365.17 USD
100208-812 100201-178 100208-810
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene ≥98%
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene

BPE, DUPHOS, Phosphine

Formula: C₁₈H₂₈P₂
MW: 306.37 g/mol
Melting Pt: 82…84 °C
MDL Number: MFCD00142336
CAS Number: 147253-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR