1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(+)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)benzene, (S,S)-Me-DUPHOS, 1,2-Bis((2S,5S)-2,5-dimethylphospholan-1-yl)benzene
15-0092-500MG
15-0092-100MG
15-0092-2G
100208-808EA
406.22
USD
100208-808
100208-806
100201-176
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene ≥98%
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene
BPE, DUPHOS, Phosphine
Formula:
C₁₈H₂₈P₂ MW: 306.37 g/mol Melting Pt: 82…84 °C |
MDL Number:
MFCD00142322 CAS Number: 136735-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...