1,2-Bis(dimethoxyphosphoryl)benzene ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tetramethyl 1,2-phenylenebis(phosphonate)

15-0089-5G 15-0089-25G
100200-852EA 171.26 USD
100200-852 100208-318
1,2-Bis(dimethoxyphosphoryl)benzene ≥99%
1,2-Bis(dimethoxyphosphoryl)benzene

Phosphine

Formula: C₁₀H₁₆O₆P₂
MW: 294.18 g/mol
Melting Pt: 80…82 °C
MDL Number: MFCD00142461
CAS Number: 15104-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR