Bis(dimethylamino)phosphoryl chloride ≥95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Phosphoric Acid Bis(dimethylamido) Chloride, N,N,N',N'-Tetramethylphosphorodiamidic Chloride, Bis(dimethylamino)phosphorochloridate, Bis(dimethylamido)phosphoric chloride
15-0087-50G
15-0087-10G
100202-934EA
336.86
USD
100202-934
100200-412
Bis(dimethylamino)phosphoryl chloride ≥95%
Bis(dimethylamino)phosphoryl chloride
Phosphine
Formula:
C₄H₁₂ClN₂OP MW: 170.58 g/mol Boiling Pt: 85 °C (0,35 mmHg) Density: 1,17 Flash Pt: >230 °F |
MDL Number:
MFCD00008302 CAS Number: 1605-65-8 UN: 1760 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...