Bis(diethylamino)chlorophosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

15-0084-1G 15-0084-5G
100197-384EA 66.52 USD
100197-384 100197-386
Bis(diethylamino)chlorophosphine ≥97%
Bis(diethylamino)chlorophosphine

Phosphine

Formula: C₈H₂₀ClN₂P
MW: 210.69 g/mol
Boiling Pt: 124…125 °C (15 mmHg)
Density: 1.002
MDL Number: MFCD00015493
CAS Number: 685-83-6
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR