1,2-Bis(dichlorophosphino)ethane ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

15-0082-2G 15-0082-10G 15-0082-50G
100201-444EA 142.95 USD
100201-444 100204-508 100209-306
1,2-Bis(dichlorophosphino)ethane ≥97%
1,2-Bis(dichlorophosphino)ethane

Phosphine

Formula: C₂H₄Cl₄P₂
MW: 231.813 g/mol
Boiling Pt: 70 °C (1 mmHg)
Density: 1.536
MDL Number: MFCD00009609
CAS Number: 28240-69-9
UN: 1760
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR