Bis(2-cyanoethyl)phenylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-0075-5G 15-0075-1G
100204-916EA 278.83 USD
100204-916 100208-316
Bis(2-cyanoethyl)phenylphosphine ≥97%
Bis(2-cyanoethyl)phenylphosphine

Phosphine

Formula: C₁₂H₁₃N₂P
Boiling Pt: 176…178 °C (0.5 mmHg)
Melting Pt: 72…73 °C
MDL Number: MFCD00013827
CAS Number: 15909-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR