α,α'-Bis(di-t-butylphosphino)-o-xylene ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1,2-Bis(di-tert-butylphosphinomethyl)benzene, 1,2-Bis((di-tert-butylphosphino)methyl)benzene

15-0072-500MG 15-0072-2G
100210-990EA 138.71 USD
100210-990 100199-762
α,α'-Bis(di-t-butylphosphino)-o-xylene ≥97%
α,α'-Bis(di-t-butylphosphino)-o-xylene
Formula: C₂₄H₄₄P₂
MW: 394.56 g/mol
Melting Pt: 59…62 °C
MDL Number: MFCD03094573
CAS Number: 121954-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR