1,3-Bis(di-t-butylphosphinomethyl)benzene ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

15-0065-1G 15-0065-250MG
101324-554EA 290.15 USD
101324-554 101324-552
1,3-Bis(di-t-butylphosphinomethyl)benzene ≥99%
1,3-Bis(di-t-butylphosphinomethyl)benzene

Phosphine, Pincer Ligands and Complexes

MDL Number: MFCD01630850
CAS Number: 149968-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR