(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(S)-(+)-1,1'-Binaphthalene-2,2'-diyl-phosphate, (11bS)-4-Hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide
15-0053-1G
15-0053-5G
101418-486EA
75.02
USD
101418-486
101418-488
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥98%
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate
Formula:
C₂₀H₁₃O₄P MW: 348.289 g/mol |
MDL Number:
MFCD00010045 CAS Number: 35193-64-7 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...