(S)-(-)-2,2'-Bis[di(3,5-di-t-butylphenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
15-0045-100MG
15-0045-500MG
101418-478EA
140.12
USD
101418-478
101418-480
(S)-(-)-2,2'-Bis[di(3,5-di-t-butylphenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl ≥97%
(S)-(-)-2,2'-Bis[di(3,5-di-t-butylphenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl
BIPHEP, Phosphine
Formula:
C₇₀H₉₆O₂P₂ |
MDL Number:
MFCD09753001 CAS Number: 167709-31-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...