Tris(trimethylsilyl)methane ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

14-9000-5G 14-9000-1G
100201-436EA 825.17 USD
100201-436 100196-048
Tris(trimethylsilyl)methane ≥97%
Tris(trimethylsilyl)methane
Formula: C₁₀H₂₈Si₃
Boiling Pt: 104 °C (20 mmHg)
Density: 0,827
Flash Pt: 170 °F
MDL Number: MFCD00008273
CAS Number: 1068-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR