Tris(dimethylamino)silane ≥99% (99.999%-Si), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

14-8750-25G 14-8750-5G
100209-660EA 307.14 USD
100209-660 100209-662
Tris(dimethylamino)silane ≥99% (99.999%-Si), PURATREM

Si, Silanes & Silanols for CVD & ALD, Volatile Precursors for CVD

Boiling Pt: 145…148 °C/ 4 °C (16 mmHg)
Density: 0,84
Flash Pt: 77 °F
MDL Number: MFCD00048006
CAS Number: 15112-89-7
UN: 3129
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR