5-(Trimethylsilyl)-1,2,3,4,5-pentamethyl-1,3-cyclopentadiene

Supplier: STREM CHEMICALS INC MS
Danger

14-8000-1G 14-8000-5G
100204-500EA 133.05 USD
100204-500 100201-174
5-(Trimethylsilyl)-1,2,3,4,5-pentamethyl-1,3-cyclopentadiene
5-(Trimethylsilyl)-1,2,3,4,5-pentamethyl-1,3-cyclopentadiene

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₃H₂₄Si
MW: 208.42 g/mol
Boiling Pt: 100 °C (10 mmHg)
Density: 0,833
MDL Number: MFCD00151193
CAS Number: 87778-95-8
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR